missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533562
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Marmoset, Mouse, Rat, Elephant, Panda, Bovine, Dog, Rabbit, Opossum (100%) Lizard (94%) Monkey (81%).
Members of the Frizzled family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. Frizzled 8 is highly expressed in two human cancer cell lines, indicating that it may play a role in several types of cancer. Its expression has been reported in brain, heart, kidney, pancreas, and skeletal muscle. ESTs have been isolated from embryo, eye, head/neck, heart, kidney, liver/spleen, pancreas, prostate, and testis libraries, as well as many tumor libraries. The crystal structure of the extracellular cysteine-rich domain of a similar mouse protein has been determined.Specifications
FZD8 | |
Polyclonal | |
Unconjugated | |
Fzd8 | |
frizzled 8, seven transmembrane spanning receptor; frizzled class receptor 8; frizzled family receptor 8; frizzled homolog 8; frizzled homolog 8 (Drosophila); frizzled-8; Fz8; Fz-8; Fzd8; hFZ8; HGNC:4046 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103352372, 14370, 364754, 616913, 8325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q498S8, Q61091, Q9H461 | |
Fzd8 | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human FZD8 / Frizzled 8. | |
50 μg | |
Primary | |
Bovine, Human, Mouse, Non-human Primate, Rabbit, Rat | |
Antibody | |
IgG |