missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ G-5 Supplement (100X)
G-5 Supplement (100X)
Brand: Gibco™ 17503012
37.73 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00100kg
Description
G-5 Supplement is a chemically defined, serum-free supplement based on Bottenstein’s G-5 formulation. G-5 Supplement is intended for use with Neurobasal™ media.G-5 Supplement is recommended for growth and expression of glial cells (normal and tumor) of astrocytic phenotype (astrocytes). G-5 Supplement can be used as a substitute for Bottenstein’s G-2 formulation. G-5 Supplement is provided as a 100X concentrate.
G-5 Supplement may be used with Neurobasal™ media, DMEM, DMEM⁄F-12, or other basal media..
Learn more about G-5 Supplement formulation.
Learn more about neural cell culture products from Invitrogen at www.invitrogen.com/neuralculture.
For Research Use Only. Not intended for any animal or human therapeutic of diagnostic use.
Specifications
Store in freezer (-5 to -20°C) and protect from light. |
|
Serum-free | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Sterile | |
Serum-Free |
Glial Cells | |
100 X | |
Liquid | |
Serum-Free | |
Dry Ice | |
Endotoxin, Performance, Sterility |
For Research Use Only. Not for use in diagnostic procedures.