missing translation for 'onlineSavingsMsg'
Learn More

G3BP1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E4A2, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen G3BP1
Clone 1E4A2
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16868405
View Documents
Proteintech
66486-1-IG-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16858405
View Documents
Proteintech
66486-1-IG-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen
Specifications

Specifications

G3BP1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
G3BP, G3BP 1, G3BP1, HDH VIII
G3bp1
IgG1
Protein G
Antibody
1E4A2
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
Unconjugated
Mouse
Mouse, Rat, Human, Porcine
P97855, Q13283
100513766, 10146, 171092, 27041
G3BP1 Fusion Protein ag3728
Primary
-20°C
G3BP1
Videos
SDS
Documents

Documents

Product Certifications