Learn More
G3BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13057-2-AP-150UL
 View more versions of this product
Product Code. 16831084
  • £349.00 / 150µL
Explore more special offers
Description
Description
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves c-myc mRNA preferentially at the 3′UTR. ATP- and magnesium-dependen
Specifications
Specifications
G3BP1
Polyclonal
Unconjugated
G3BP1
G3BP, G3BP 1, G3BP1, HDH VIII
Rabbit
Antigen Affinity Chromatography
RUO
10146, 171092, 27041
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97855, Q13283
G3bp1
G3BP1 Fusion Protein Ag3728
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
G3BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated