missing translation for 'onlineSavingsMsg'
Learn More

G418 disulfate salt, Tocris Bioscience™

Product Code. 18095852
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aminoglycoside antibiotic; used in cell culture

G418 disulfate salt is an aminoglycosidic antibiotic. Inhibits protein synthesis in both prokaryotic and eukaryotic cells. Used for selection of eukaryotic cells transfected with the neo gene.

This product has been cell culture tested.

Specifications

Type G418 Disulfate
CAS 108321-42-2
Quantity 100 mg
Molecular Formula C20H44N4O18S2
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Molecular Weight (g/mol) 692.70
PubChem CID 134129582
Formula Weight 692.71
Percent Purity >98%
Chemical Name or Material G418 disulfate salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'