missing translation for 'onlineSavingsMsg'
Learn More

G418 Sulfate (White to Off-white Powder), Fisher BioReagents

£110.00 - £309.00

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
PubChem CID 134129582
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10204773
View Documents Promotion Details
Fisher BioReagents
BP673-1
1 g Amber Glass
£110.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10648393
View Documents Promotion Details
Fisher BioReagents
BP673-5
5 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

108321-42-2
692.70
UHEPSJJJMTWUCP-KETIWVBHNA-N
(2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
C20H44N4O18S2
MFCD00058314
134129582
OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

G418 Sulfate
< 0.015 at 280nm, < 0.10 at 570nm
> 98 %
MFCD00058314
UHEPSJJJMTWUCP-KETIWVBHNA-N
(2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
134129582
>98%
White
Solid
108321-42-2
C20H44N4O18S2
> 730μg/mg
OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
692.70
692.7
10% Max.
G418 Sulfate
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : -20°C

Product Identifier
  • G418 sulfate

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1A
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS
Documents

Documents

Product Certifications