missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
GABA is a GABA receptor agonist and endogenous inhibitory neurotransmitter. Caged RuBi GABA is also available (Cat. No. 3400).
Specifications
Specifications
CAS | 56-12-2 |
Molecular Formula | C4H9NO2 |
Synonym | 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino |
InChI Key | BTCSSZJGUNDROE-UHFFFAOYSA-N |
SMILES | C(CC(=O)O)CN |
IUPAC Name | 4-aminobutanoic acid |
Molecular Weight (g/mol) | 103.121 |
PubChem CID | 119 |
ChEBI | CHEBI:16865 |
Formula Weight | 103.12 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction