missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABARAPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11010-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to GABARAPL1, and does not recognize the recombit GABARAP and GABARAPL2.
Increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.Specifications
GABARAPL1 | |
Polyclonal | |
Unconjugated | |
GABARAPL1 | |
APG8 LIKE, APG8L, ATG8, ATG8L, GABARAPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23710, 57436, 689161 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q0VGK0, Q8R3R8, Q9H0R8 | |
Gabarapl1 | |
GABARAPL1-Specific Fusion Protein Ag1473 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |