missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14104-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.Specifications
GABRG2 | |
Polyclonal | |
Unconjugated | |
GABRG2 | |
CAE2, ECA2, GABA A receptor gamma 2, GABAA receptor gamma 2, GABRG2, GEFSP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14406, 2566, 29709 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18507, P18508, P22723 | |
Gabrg2 | |
GABRG2 Fusion Protein Ag5237 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |