Learn More
GABRP (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111781
Additional Details : Weight : 0.01000kg
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)DGNDVEF SWLRGND Homology: Mouse amino acid residues identical; human - 13/14 amino acid residues identical.
GABRP (extracellular) Polyclonal antibody specifically detects GABRP (extracellular) in Mouse, Rat samples. It is validated for Western BlotSpecifications
GABRP (extracellular) | |
Polyclonal | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O09028, Q8QZW7 | |
Rabbit | |
Affinity Purified | |
RUO | |
216643, 81658 | |
-20°C | |
Lyophilized |
Western Blot | |
Unconjugated | |
GABRP | |
GABA(A) receptor subunit pi; GABA(A) receptor, pi; GABA-A receptor pi subunit; GABRP; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, pi; gamma-aminobutyric acid (GABA-A) receptor, pi; gamma-aminobutyric acid A receptor, pi; gamma-aminobutyric acid receptor subunit pi; gamma-aminobutyric acid type A receptor pi subunit; MGC126386; MGC126387 | |
Peptide corresponding to amino acid residues 184-197 of rat GABA(A) pi Receptor (Extracellular, N-terminus) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |