missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABRR3 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577412
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.Specifications
GABRR3 (extracellular) | |
Polyclonal | |
Unconjugated | |
Gabrr3 | |
GABA(A) rho3 Receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
192258, 328699 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P50573 | |
Gabrr3 | |
Peptide CLSSPKQTRIRETR, corresponding to amino acid residues 31-44 of rat GABA(A) rho3 Receptor | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |