missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium boride, 99.5% (metals basis)

Product Code. 11303628
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11303628

Brand: Thermo Scientific Alfa Aesar 039225.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12008-06-9 | B6Gd | 222.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-06-9
Molecular Formula B6Gd
Molecular Weight (g/mol) 222.11
MDL Number MFCD00049942
InChI Key FYKPSYVUFHLGOY-UHFFFAOYSA-N
Synonym gadolinium boride, gadolinium boride, gdb6, gadolinium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide, gadolinium 2+ ion pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide
PubChem CID 25021694
IUPAC Name gadolinium(2+) pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
SMILES [Gd++].B12B3[B-]11B4B1[B-]234

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 222.12
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Gadolinium boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.