missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11386648

Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11386648
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11386648

Brand: Thermo Scientific Alfa Aesar 013204.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64438-54-6
Molecular Formula C15H21GdO6
Molecular Weight (g/mol) 454.58
MDL Number MFCD00013493
InChI Key PJCXQIZIMGZZIT-UHFFFAOYSA-N
Synonym gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
PubChem CID 131675701
IUPAC Name gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
SMILES [Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 454.58 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Gadolinium(III) 2,4-pentanedionate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN06J074 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number013204.14,013204.06
Lot NumberK26T018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013204.14,013204.06
Lot NumberL27X030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013204.14,013204.06
Lot NumberH28T011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013204.14,013204.06
Lot NumberL23Z032 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013204.14,013204.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.