missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) carbonate hydrate, 99% (REO), Thermo Scientific Chemicals Product Code.: 11303778

Gadolinium(III) carbonate hydrate, 99% (REO), Thermo Scientific Chemicals

Product Code. 11303778
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11303778

Brand: Thermo Scientific Alfa Aesar 040254.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-36-2
Molecular Formula C3Gd2O9
Molecular Weight (g/mol) 494.52
MDL Number MFCD00150240
InChI Key RQXZRSYWGRRGCD-UHFFFAOYSA-H
Synonym gadolinium carbonate hydrate, gadolinium iii carbonate hydrate, acmc-1aeff, 3co3.2gd.h2o, gadolinium carbonate-water 2/3/1, digadolinium 3+ hydrate tricarbonate, gadolinium iii carbonate hydrate, reacton?, gadolinium iii carbonate hydrate-gd reo puratrem
PubChem CID 16212943
SMILES [Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 5 g
IUPAC Name gadolinium(3+);tricarbonate;hydrate
Formula Weight 494.53 (Anhydrous)
Percent Purity 99%
Assay (REO)
Chemical Name or Material Gadolinium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) carbonate hydrate, 99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.