missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Gadolinium(III) carbonate is used for making optical glass and dopant for Gadolinium Yttrium Garnets which have microwave applications. It is also used for making phosphors for color TV tube and energy-saving lamps. It is used as intravenous MRI contrast agent to enhance images in medical magnetic resonance imaging and Magnetic Resonance Angiography (MRA) procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Formula Weight | 494.53 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Assay | (REO) |
Chemical Name or Material | Gadolinium(III) carbonate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'