missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Product Code. 11391009
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11391009

Brand: Thermo Scientific Alfa Aesar 011287.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 13450-84-5 | Cl3GdH12O6 | 371.69 g/mol

Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-84-5
Molecular Formula Cl3GdH12O6
Molecular Weight (g/mol) 371.69
MDL Number MFCD00149732
InChI Key PNYPSKHTTCTAMD-UHFFFAOYSA-K
Synonym gadolinium chloride hexahydrate, gadolinium trichloride hexahydrate, unii-261ex557hi, gadolinium chloride, hydrate 1:6, gadolinium chloride gdcl3 , hexahydrate, gadolinium chloride gdcl3 , hexahydrate 8ci,9ci, gadolinium 3+ trichloride hexahydrate, gadolinium iii chloridehexahydrate, gadolinium chloride, hexahydrate,, gadolinium iii chloride hexahydrate, reacton reo
PubChem CID 197915
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 5 g
Merck Index 14,4326
Solubility Information soluble in water.
IUPAC Name gadolinium(3+);trichloride;hexahydrate
Formula Weight 371.70 (263.61 Anhydrous)
Percent Purity 99.99%
Grade Reacton
Odor Characteristic
Sensitivity Hygroscopic
Density 2.424 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Gadolinium(III) chloride hexahydrate, REacton
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Gadolinium chloride(GdCl3), hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.