Learn More
Gadolinium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
£149.75 - £531.65
Chemical Identifiers
CAS | 10138-52-0 |
---|---|
Molecular Formula | Cl3Gd |
Molecular Weight (g/mol) | 263.6 |
MDL Number | MFCD00011024 |
InChI Key | MEANOSLIBWSCIT-UHFFFAOYSA-K |
Synonym | gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3Show More |
PubChem CID | 61486 |
ChEBI | CHEBI:37288 |
IUPAC Name | trichlorogadolinium |
SMILES | Cl[Gd](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358607
|
Thermo Scientific Alfa Aesar
035736.06 |
5 g |
£149.75
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348607
|
Thermo Scientific Alfa Aesar
035736.14 |
25 g |
£531.65
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gadolinium(III) Chloride is used as chelating agents, which is essential for biomedical applications. It is also employed in the synthesis and nuclear magnetic resonance (NMR) studies of paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes. It plays an important role as contrast agent for magnetic resonance imaging and an macrophage inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10138-52-0 | |
263.6 | |
MEANOSLIBWSCIT-UHFFFAOYSA-K | |
61486 | |
trichlorogadolinium |
Cl3Gd | |
MFCD00011024 | |
gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3Show More | |
CHEBI:37288 | |
Cl[Gd](Cl)Cl |
Specifications
609°C | |
White | |
99.99% (REO) | |
MFCD00011024 | |
Soluble in water. | |
Cl[Gd](Cl)Cl | |
263.6 | |
CHEBI:37288 | |
Hygroscopic | |
4.52g/cm3 |
10138-52-0 | |
Powder | |
Cl3Gd | |
gadolinium chloride, gadolinium trichloride, gadolinium chloride gdcl3, gdcl3, gadolinium iii chloride, gadolinium 3+ chloride, wln: gd g3, gadolinium iii chloride, ultra dry, gadolinium iii chloride, anhydrous, ?gdcl3 | |
MEANOSLIBWSCIT-UHFFFAOYSA-K | |
trichlorogadolinium | |
61486 | |
263.61 | |
White powder | |
Gadolinium(III) chloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
missing translation for 'einecsNumber' : 233-386-6
missing translation for 'rtecsNumber' : LW4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.