missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11346908

Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11346908
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11346908

Brand: Thermo Scientific Alfa Aesar 013655.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-26-9
Molecular Formula F3Gd
Molecular Weight (g/mol) 214.25
MDL Number MFCD00016113
InChI Key TYIZUJNEZNBXRS-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trifluoride
SMILES [F-].[F-].[F-].[Gd+3]

Specifications

Melting Point 1231°C
Color White
Physical Form Crystalline
Quantity 100 g
Formula Weight 214.25
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 7.1 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) fluoride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL26Z013 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number013655.14,013655.22
Lot NumberF16Q06 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number013655.14,013655.22
Lot NumberN01D025 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number013655.14,013655.22
Lot NumberH21T013 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number013655.14,013655.22
Lot NumberM13C050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number013655.14,013655.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.