missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11321029

Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11321029
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11321029

Brand: Thermo Scientific Alfa Aesar 011293.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-26-9
Molecular Formula F3Gd
Molecular Weight (g/mol) 214.25
MDL Number MFCD00016113
InChI Key TYIZUJNEZNBXRS-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trifluoride
SMILES [F-].[F-].[F-].[Gd+3]

Specifications

Melting Point 1231°C
Color White
Physical Form Crystalline
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 214.25
Percent Purity 99.99%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 7.1 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.