missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11397647

Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11397647
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 11397647

Brand: Thermo Scientific Alfa Aesar 012917.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94219-55-3
Molecular Formula GdN3O9
Molecular Weight (g/mol) 343.26
MDL Number MFCD00011023
InChI Key MWFSXYMZCVAQCC-UHFFFAOYSA-N
IUPAC Name gadolinium(3+) trinitrate
SMILES [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 92°C
Color White
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,4326
Solubility Information Soluble in water.
Formula Weight 343.26 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 2.406 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) nitrate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ06E008 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number012917.14,012917.22
Lot NumberN22J017 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number012917.14,012917.22
Lot NumberY16H028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012917.14,012917.22
Lot NumberH07K13 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012917.14,012917.22
Lot Number61300570 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012917.14,012917.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) nitrate hydrate, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.