missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate pentahydrate, 99.9%, Thermo Scientific Chemicals

Product Code. 10246590
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52788-53-1
Molecular Formula GdN3O9·5H2O
Molecular Weight (g/mol) 433.33
MDL Number MFCD00149729

Specifications

Melting Point 92.0°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Linear Formula Gd(NO3)3·5H2O
Merck Index 14, 4326
Formula Weight 433.33
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate pentahydrate, 99.9%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gadolinium(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'