missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitrate pentahydrate, 99.9%

Product Code. p-222420 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10654712

Brand: Thermo Scientific Acros 199140250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 52788-53-1 | GdN3O9·5H2O | 433.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52788-53-1
Molecular Formula GdN3O9·5H2O
Molecular Weight (g/mol) 433.33
MDL Number MFCD00149729

Specifications

Melting Point 92.0°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9%
Linear Formula Gd(NO3)3·5H2O
Merck Index 14, 4326
Formula Weight 433.33
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate pentahydrate, 99.9%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Gadolinium(III) nitrate pentahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.