missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 15472747

Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 15472747
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15472747

Brand: Thermo Scientific Alfa Aesar 046709.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25764-15-2
Molecular Formula GdN
Molecular Weight (g/mol) 171.26
MDL Number MFCD00049943
InChI Key FLATXDRVRRDFBZ-UHFFFAOYSA-N
Synonym gadolinium nitride, gadolinium iii nitride, gadoliniumylidyneamine
PubChem CID 117632
IUPAC Name azanylidynegadolinium
SMILES N#[Gd]

Specifications

Color Black
Physical Form Powder
Quantity 5 g
Formula Weight 171.26
Percent Purity 99.5%
Assay (REO)
Chemical Name or Material Gadolinium (III) nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.