missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) oxalate hydrate, 99.9% (REO), Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Conditionnement:
100g
25g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Spécification
| Boiling Point | (decomposition) |
| Melting Point | 110°C (in -6H2O) |
| Color | White |
| Physical Form | Powder |
| Quantity | 25 g |
| Linear Formula | Gd2(C2O4)3·xH2O |
| UN Number | UN3288 |
| Formula Weight | 578.56 (Anhydrous) |
| Percent Purity | 99.9% |
| Sensitivity | Hygroscopic |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu