missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, nanopowder, 99.99+% (REO)

Product Code. 11335098
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11335098

Brand: Thermo Scientific Alfa Aesar 044020.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption materials in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
IUPAC Name digadolinium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 2330°C
Physical Form Powder
Quantity 500 g
Merck Index 14,4326
Solubility Information Soluble in acid. Insoluble in water.
Formula Weight 362.5
Percent Purity ≥99.99%
Density 7.407 g/mL
Assay (REO)
Chemical Name or Material Gadolinium (III) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.