Learn More
Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
£26.63 - £266.33
Chemical Identifiers
CAS | 14017-52-8 |
---|---|
MDL Number | MFCD00150239 |
Synonym | gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2oShow More |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470291
|
Thermo Scientific Alfa Aesar
040571.NA |
2 g |
£26.63
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480291
|
Thermo Scientific Alfa Aesar
040571.NC |
10 g |
£78.59
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490291
|
Thermo Scientific Alfa Aesar
040571.NE |
50 g |
£266.33
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14017-52-8 | |
MFCD00150239 | |
gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o |
Specifications
14017-52-8 | |
MFCD00150239 | |
gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o | |
563.69 (455.60 Anhydrous) | |
50% w/w aq. soln. | |
Gadolinium(III) perchlorate hexahydrate |
Liquid | |
UN3211 | |
Insoluble in water. | |
Reagent | |
Odorless |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
missing translation for 'einecsNumber' : 237-837-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.