missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11360108

Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11360108
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11360108

Brand: Thermo Scientific Alfa Aesar 042869.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cryogenic research

Gadolinium(III) sulfate is used as analytical reagents, in inorganic synthesis and electronic ceramics. It is also useful in cryogenic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-87-8
Molecular Formula Gd2H16O20S3
Molecular Weight (g/mol) 746.788
MDL Number MFCD00149734
InChI Key JBFCVGZGWFIPNK-UHFFFAOYSA-H
Synonym gadolinium iii sulfate octahydrate, digadolinium 3+ octahydrate trisulfate, gadolinium iii sulfate, gadolinium sulfate octahydrate, gadolinium sulfuric acid octahydrate, gadolinium sulfate, gadolinium sulfate-water 2/3/8, gadolinium iii sulfate octahydrate, reacton, gadolinium iii sulfate octahydrate trace metals basisShow MoreShow Less
PubChem CID 16211473
IUPAC Name gadolinium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Gd+3].[Gd+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Merck Index 14,4326
Solubility Information Soluble in cold water
Formula Weight 746.81 (602.68 Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Density 4.139 g/mL
Assay (REO)
Chemical Name or Material Gadolinium (III) sulfate octahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253631 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number042869.18,042869.09
Lot NumberZ27K028 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number042869.18,042869.09
Lot NumberQ29F019 Certificate TypeCertificate of Analysis Date15/04/2022 Catalog Number042869.18,042869.09
Lot NumberX28H016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number042869.18,042869.09
Lot NumberI16Z024 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number042869.18,042869.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium(III) sulfate octahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.