missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15482557
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Color Silver
UN Number UN3089
Quantity 5 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Gadolinium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'