missing translation for 'onlineSavingsMsg'
Learn More
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals Product Code.: 11303828

Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Product Code. 11303828
10 mm
Click to view available options
Quantity:
10 mm
25 mm
This item is not returnable. View return policy

Product Code. 11303828

Brand: Thermo Scientific Alfa Aesar 040290.HF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Quantity 10 mm
Percent Purity 99.9%
Packaging Mylar under Argon
Weight ≈0.97g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Gadolinium rod, 12.7mm (0.5 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR05I002 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog Number040290.HF,040290.DH
Lot NumberE12K22 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040290.HF,040290.DH
Lot NumberC13N11 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040290.HF,040290.DH
Lot NumberB01T026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040290.HF,040290.DH
Lot NumberP11F052 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040290.HF,040290.DH

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gadolinium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.