missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium solution for ICP 1g/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£86.85 - £127.00
Specifications
Concentration | 1±0.02g/L |
---|---|
Type | Mass Spectrometry Standard |
Analyte or Component Names | Gd |
Solvent or Matrix | HNO3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12903414
|
Fisher Chemical
S 021 100 |
100 mL | Bottle |
£92.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12913414
|
Fisher Chemical
S 021 250 |
250 mL | Bottle |
£86.85
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12923414
|
Fisher Chemical
S 021 500 |
500 mL | Bottle |
£127.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1±0.02g/L | |
Mass Spectrometry Standard | |
Gd | |
HNO3 |
Safety and Handling
H225; H304; H315; H336; H410
P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501
Danger
missing translation for 'shelfLife' : 24 months (12 months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title