missing translation for 'onlineSavingsMsg'
Learn More
β-Galactosidase, MP Biomedicals™ Product Code.: 11486432

β-Galactosidase, MP Biomedicals™

Product Code. 11486432 Shop All MP Biomedicals Products
3 kU
Click to view available options
Quantity:
3 kU
This item is not returnable. View return policy

Product Code. 11486432

Brand: MP Biomedicals 0215003903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 9041-08-1
Color White
MDL Number MFCD00081689
Quantity 3 kU
Synonym Lactase, β-D-Galactoside galactohydrolase
Physical Form Powder
Chemical Name or Material β-Galactosidase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
β-Galactosidase, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.