missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-13 Mouse, Clone: PP13/1165, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 29124-MSM5-P0
This item is not returnable.
View return policy
Specifications
Galectin-13 | |
Cytoplasmic and Cell Surface | |
PP13/1165 | |
Unconjugated | |
XP_016882204 | |
Recombinant human Galectin-13 protein fragment (aa23-134) | |
K-562 cells. Placenta or Spleen. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Mouse | |
16kDa (monomer) ;32kDa (homodimer) | |
Protein A/G Purified | |
RUO | |
29124 | |
IgG2b |