Learn More
Galena, naturally occurring mineral, approximately 1-2in, Thermo Scientific Chemicals
£20.40 - £70.10
Chemical Identifiers
CAS | 1314-87-0 |
---|---|
Molecular Formula | PbS |
Molecular Weight (g/mol) | 239.30 |
MDL Number | MFCD00016280 |
InChI Key | FILXVLQXZYXBSJ-UHFFFAOYSA-N |
Synonym | lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide |
PubChem CID | 14819 |
IUPAC Name | sulfanylidenelead |
SMILES | [S--].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466485
|
Thermo Scientific Alfa Aesar
042546.KF |
1 Ea. |
£20.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15476485
|
Thermo Scientific Alfa Aesar
042546.KJ |
5 Pc. |
£70.10
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-87-0 | |
239.30 | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
14819 | |
[S--].[Pb++] |
PbS | |
MFCD00016280 | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
sulfanylidenelead |
Specifications
1314-87-0 | |
PbS | |
UN3077 | |
Soluble in strong HNO3,in excess of hot HCl | |
[S--].[Pb++] | |
239.30 | |
239.25 | |
Galena |
Crystal Clusters | |
MFCD00016280 | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
sulfanylidenelead | |
14819 | |
Bag |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 215-246-6
missing translation for 'rtecsNumber' : OG4550000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only