Learn More
Thermo Scientific™ Gallery™ System Reagents for Environmental and Industrial Analysis
Choose from a continuously expanding selection of tests for automated water quality monitoring and nutrient analysis.
Brand: Thermo Scientific™ 984631
Description
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery™ system reagents for environmental analysis.
Highlights:- All test applications are based on recommended standard methods in water and wastewater analysis of different countries
- Kit structure includes 1 to 3 reagents in 20mL vials
- Barcoded vials are simple and fast to use
- All necessary data, such as lot and expiration date, are automatically read
- Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
- Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
Fluoride System Reagent | |
Fluoride system reagents - R3 (2200 tests) | |
2200 | |
4 x 20 mL |
Water Sample, Soil Sample | |
Fluoride R3 | |
0 to 5.0 (for Gallery), 0 to 2.0 (for Aquakem) | |
600/ 880 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.