missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid 98%, Thermo Scientific Chemicals
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 253.0°C |
Color | Yellow to Beige |
Assay Percent Range | 98% |
Solubility Information | Solubility in water: 12g/L cold water. Other solubilities: easily soluble in warmed water, very soluble in ethanol: 160g/L, acetone: 200g/l, soluble in glycerol: 100g/L, slightly soluble in, ether: 10g/L, insoluble in benzene, chloroform, petrol ether |
Formula Weight | 170.12 |
Percent Purity | 98% |
Density | 1.6940g/mL |
Quantity | 5 g |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Gallic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'