missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
£28.94 - £307.53
Chemical Identifiers
CAS | 7440-55-3 |
---|---|
Molecular Formula | Ga |
Molecular Weight (g/mol) | 69.72 |
MDL Number | MFCD00134045 |
InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
Synonym | gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom |
PubChem CID | 5360835 |
ChEBI | CHEBI:49631 |
IUPAC Name | gallium |
SMILES | [Ga] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10328670
|
Thermo Scientific Acros
193070010 |
1 g | Plastic bottle |
£28.94
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246920
|
Thermo Scientific Acros
193070050 |
5 g | Plastic bottle |
£89.34
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924081
|
Thermo Scientific Acros
193070250 |
25 g | Plastic bottle |
£307.53
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
5360835 | |
gallium |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
CHEBI:49631 | |
[Ga] |
Specifications
Gray | |
29.7°C | |
5.9000g/mL | |
Ga | |
MFCD00134045 | |
Solubility in water: insoluble. | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
99.9999% | |
Solid and/or Liquid |
7440-55-3 | |
2400.0°C | |
99.9999% | |
Ga | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
69.72 | |
Trace Metal Basis | |
Gallium |
Safety and Handling
Warning
Recommended Storage : 4°C
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Gallium
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only