Learn More
Gallium antimonide, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Gallium antimonide can be used for Infrared detectors, infrared LEDs and lasers and transistors, and thermophotovoltaic systems. It can be used as a component of photoresists and other composites where high refractive index is desirable
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 710°C |
Physical Form | Pieces |
Quantity | 5 g |
UN Number | UN1549 |
Solubility Information | Insoluble in water. |
Formula Weight | 191.47 |
Percent Purity | 99.99% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Gallium antimonide |
Suggestions de produits
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M11H076 | Certificate of Analysis | 11/06/2024 | 012933.03,012933.06 |
P17B058 | Certificate of Analysis | 24/10/2022 | 012933.03,012933.06 |
H29L01 | Certificate of Analysis | 08/12/2021 | 012933.03,012933.06 |
A22S008 | Certificate of Analysis | 08/12/2021 | 012933.03,012933.06 |
G05X055 | Certificate of Analysis | 07/12/2021 | 012933.03,012933.06 |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.