missing translation for 'onlineSavingsMsg'
Learn More

Gallium(II) telluride, 99.999% (metals basis)

Product Code. p-7051734
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11354857

Brand: Thermo Scientific Alfa Aesar 041881.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12024-14-5 | GaTe | 197.323 g/mol

Gallium(II) telluride acts as a semiconductor is used in the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-14-5
Molecular Formula GaTe
Molecular Weight (g/mol) 197.323
MDL Number MFCD00135536
InChI Key OFIYHXOOOISSDN-UHFFFAOYSA-N
Synonym gallium ii telluride
PubChem CID 14496722
IUPAC Name tellanylidenegallium
SMILES [Ga]=[Te]

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 197.32
Percent Purity 99.999%
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Gallium(II) telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.