missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11379727

Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11379727
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11379727

Brand: Thermo Scientific Alfa Aesar 042086.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14405-43-7
Molecular Formula C15H21GaO6
Molecular Weight (g/mol) 367.05
MDL Number MFCD00013492
InChI Key IMMRYVXLCAMRNK-UHFFFAOYSA-N
Synonym gallium iii acetylacetonate, gallium iii 2,4-pentanedionate
PubChem CID 71311502
IUPAC Name (Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one
SMILES [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 196°C to 198°C (decomposition)
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 367.05
Percent Purity 99.99%
Odor Odorless
Density 1.42 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) 2,4-pentanedionate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ12K003 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number042086.14,042086.06
Lot NumberX03K052 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number042086.14,042086.06
Lot NumberZ21J005 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number042086.14,042086.06
Lot NumberU28J026 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number042086.14,042086.06
Lot NumberX31I038 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog Number042086.14,042086.06
compliance-icons compliance-icons
Product Identifier
  • Gallium(III) 2,4-pentanedionate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Gallium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) 2,4-pentanedionate, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.