missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) bromide, anhydrous, Thermo Scientific Chemicals Product Code.: 11368327

Gallium(III) bromide, anhydrous, Thermo Scientific Chemicals

Product Code. 11368327
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11368327

Brand: Thermo Scientific Alfa Aesar 032107.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gallium(III) bromide is used as organic intermediates. It is a powerful oxidizing agent and is able to release oxygen free radicals from the water in mucous membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-88-9
Molecular Formula Br3Ga
Molecular Weight (g/mol) 309.435
MDL Number MFCD00016100
InChI Key SRVXDMYFQIODQI-UHFFFAOYSA-K
Synonym gallium tribromide, gallium bromide gabr3, gallium iii bromide, gallium bromide, gabr3, gallium bromide, gabr3, gallium iii bromide, anhydrous, gallium iii bromide, ultra dry, gallium iii bromide, anhydrous, granular
PubChem CID 83477
IUPAC Name tribromogallane
SMILES [Ga](Br)(Br)Br

Specifications

Boiling Point 279°C
Melting Point 122°C
Color White
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Solubility Information Slightly soluble in water. Soluble in acids.
Formula Weight 309.45
Odor Odorless
Sensitivity Moisture sensitive
Density 3.69 g/mL
Chemical Name or Material Gallium(III) bromide, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ23K053 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number032107.04,032107.09
Lot NumberW25J055 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number032107.04,032107.09
Lot NumberT13J046 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number032107,032107.04,032107.09
Lot NumberX04I061 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number032107.04,032107.09
Lot NumberZ13D003 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number032107.04,032107.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) bromide, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.