missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific Chemicals Product Code.: 12048598

Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific Chemicals

Product Code. 12048598
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 12048598

Brand: Thermo Scientific Acros 444100050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.08
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliueShow MoreShow Less
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201.0°C
Melting Point 78.0°C
Quantity 5 g
Assay Percent Range 99.99+%
Linear Formula GaCl3
Merck Index 15,4378
Solubility Information Solubility in water: reacts
Formula Weight 176.08
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gallium(III) Chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463244 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number444100250,444101000,444100050
Lot NumberA0460686 Certificate TypeCertificate of Analysis Date29/02/2024 Catalog Number444100250,444101000,444100050
Lot NumberA0450799 Certificate TypeCertificate of Analysis Date25/04/2023 Catalog Number444100250,444101000,444100050
Lot NumberA0401728 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number444100250,444101000,444100050
Lot NumberA0399800 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number444100250,444101000,444100050
compliance-icons
Product Identifier
  • Gallium(III) chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402-Store in a dry place.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.