missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 15496675

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15496675
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15496675

Brand: Thermo Scientific Alfa Aesar 045026.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalyst for a number of transformations including the ring opening and recyclization of epoxides and the trimerization of alkynes. It is used to detect the solar neutrions.

Gallium(III) chloride is a catalyst for a number of transformations including the ring opening and recyclization of epoxides and the trimerization of alkynes. It is used to detect the solar neutrions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.073
MDL Number MFCD00011018
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201°C
Melting Point 78°C
Physical Form Granules
Quantity 25 g
UN Number UN3260
Solubility Information Soluble in water,benzene,carbon tetrachloride and carbon disulfide
Formula Weight 176.03
Percent Purity 99.999%
Packaging Ampouled under argon
Sensitivity Moisture sensitive
Specific Gravity 2.47
Density 2.47
Chemical Name or Material Gallium(III) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253618 Certificate TypeCertificate of Analysis Date08/01/2025 Catalog Number045026.14,045026.22,045026.06
Lot NumberY13K013 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number045026.14,045026.22,045026.06
Lot NumberZ27K040 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number045026.14,045026.22,045026.06
Lot NumberR08K005 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number045026.14,045026.22,045026.06
Lot NumberM20I042 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045026.14,045026.22,045026.06
compliance-icons
Product Identifier
  • Gallium(III) chloride, ultra dry
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Gallium trichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.