missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium(III) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Spécification
| Boiling Point | 201°C |
| Melting Point | 78°C |
| Color | White |
| Physical Form | Beads |
| Quantity | 25 g |
| Assay Percent Range | 99.999% (Metal basis) |
| Solubility Information | Very soluble in water. Soluble in benzene,carbon tetrachloride and carbon disulfide |
| Formula Weight | 176.03 |
| Sensitivity | Moisture sensitive |
| Appearance | Ultra dry. White powder/flake/crystalline/beads, etc. |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Spot an opportunity for improvement?Share a Content Correction