missing translation for 'onlineSavingsMsg'
Learn More
Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals Product Code.: 11318337

Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals

Product Code. 11318337
0.5 g
Click to view available options
Quantity:
0.5 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11318337

Brand: Thermo Scientific Alfa Aesar 032116.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Gallium(III) nitrate hydrate is used in the preparation of gallium nitride and other gallium compounds. It acts as a drug used to treat symptomatic hypercalcemia secondary to cancer. It finds application as an antineoplastic agent and also as an acting agent for the treatment of bone-resorptive diseases. It is involved to improve the cultural selectivity of medically significant fungi by suppression of contaminating bacterial flora.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69365-72-6
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
PubChem CID 11550823
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Physical Form Powder, Flake, Crystalline, Beads, Lump, Etc.
Quantity 0.5 g
Assay Percent Range 99.9% (Metal basis)
IUPAC Name gallium(3+) trinitrate
Formula Weight 255.73 (anhydrous)
Odor Odorless
Appearance White powder/flake/crystalline/beads/lump, etc.
Chemical Name or Material Gallium(III) nitrate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP19L006 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number032116.02,032116.14,032116.06
Lot NumberP19L007 Certificate TypeCertificate of Analysis Date21/03/2025 Catalog Number032116.02,032116.14,032116.06
Lot NumberM28L049 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number032116.02,032116.14,032116.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.