Learn More
Gallium (III) Nitrate Hydrate, 99.9%, metals basis, Thermo Scientific Chemicals
Description
Gallium(III) nitrate hydrate is used in the preparation of gallium nitride and other gallium compounds. It acts as a drug used to treat symptomatic hypercalcemia secondary to cancer. It finds application as an antineoplastic agent and also as an acting agent for the treatment of bone-resorptive diseases. It is involved to improve the cultural selectivity of medically significant fungi by suppression of contaminating bacterial flora.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 110°C (decomposition) |
Color | White |
Physical Form | Lump |
Quantity | 25 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | Soluble in water. |
IUPAC Name | gallium;trinitrate;hydrate |
Formula Weight | 255.73 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P19L006 | Certificate of Analysis | 21/03/2025 | 032116.02,032116.14,032116.06 |
P19L007 | Certificate of Analysis | 21/03/2025 | 032116.02,032116.14,032116.06 |
M28L049 | Certificate of Analysis | 30/01/2025 | 032116.02,032116.14,032116.06 |
S24K027 | Certificate of Analysis | 13/11/2024 | 032116.14,032116.06 |
U21F080 | Certificate of Analysis | 12/04/2022 | 032116.14,032116.06 |
Safety and Handling


- Gallium(III) nitrate hydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Gallium (III) Nitrate Hydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.