Learn More
Gallium(III) nitrate solution, Ga 9-10% w/w, Thermo Scientific Chemicals
Description
Gallium(III) nitrate solution, is used as the material for producing gallium salts and for scientific research and chemical reagent. It is also used as an intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Liquid |
Quantity | 50 g |
UN Number | UN3218 |
Solubility Information | It is soluble in water. |
Formula Weight | 255.73 |
Concentration or Composition (by Analyte or Components) | Ga 9 to 10% w/w |
Chemical Name or Material | Gallium (III) nitrate solution |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M15H018 | Certificate of Analysis | 11/11/2022 | 043484.30,043484.18 |
Y19B055 | Certificate of Analysis | 18/11/2021 | 043484.30,043484.18 |
R28G059 | Certificate of Analysis | 18/11/2021 | 043484.30,043484.18 |
D15W025 | Certificate of Analysis | 18/11/2021 | 043484.30,043484.18 |
A28M23 | Certificate of Analysis | 18/11/2021 | 043484.30,043484.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.