missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) selenide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15412377
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Gallium (III) Selenide is a crystalline solid used as a semiconductor and in photo optic applications. It is also used as an intermediate in food, agricultural and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-24-7
Molecular Formula Ga2Se3
MDL Number MFCD00016103

Specifications

Physical Form Sintered Lump
Quantity 1 g
UN Number UN3077
Solubility Information Decomposes with water.
Formula Weight 376.33
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Gallium (III) selenide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'