missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) selenide, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. p-7059900
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15412377

Brand: Thermo Scientific Alfa Aesar 045572.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gallium (III) Selenide is a crystalline solid used as a semiconductor and in photo optic applications. It is also used as an intermediate in food, agricultural and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12024-24-7
Molecular Formula Ga2Se3
MDL Number MFCD00016103

Specifications

Physical Form Sintered Lump
Quantity 1 g
UN Number UN3077
Solubility Information Decomposes with water.
Formula Weight 376.33
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Gallium (III) selenide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.