Learn More
Gallium Indium Tin eutectic, 99.99% (metals basis), Thermo Scientific Chemicals
£27.94 - £389.00
Specifications
Chemical Name or Material | Gallium Indium Tin eutectic |
---|---|
Melting Point | 10.7°C |
Color | White |
Physical Form | Liquid |
Assay Percent Range | 99.99% (Metals basis) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376948
|
Thermo Scientific Alfa Aesar
014634.03 |
1 g |
£27.94
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356948
|
Thermo Scientific Alfa Aesar
014634.09 |
10 g |
£118.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366948
|
Thermo Scientific Alfa Aesar
014634.18 |
50 g |
£389.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gallium-Indium eutectic (EGAIn) is an electrically conductive fluid metal. It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gallium Indium Tin eutectic | |
White | |
99.99% (Metals basis) | |
MFCD00144388 | |
Odorless |
10.7°C | |
Liquid | |
Ga:In:Sn; 62:22:16 wt.% | |
Insoluble in water or organic solvents. | |
6.359 |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packing Group: III
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only