missing translation for 'onlineSavingsMsg'
Learn More
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11381238

Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11381238
500 g
Click to view available options
Quantity:
50 g
500 g
This item is not returnable. View return policy

Product Code. 11381238

Brand: Thermo Scientific Alfa Aesar 010188.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atomShow MoreShow Less
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Gray-Silver
UN Number UN2803
Quantity 500 g
Percent Purity 99.99%
Form Ingot
Assay (metals basis)
Chemical Name or Material Gallium ingot

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY18K027 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog Number010188.18,010188.14,010188.36,010188.30
Lot NumberS22J028 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number010188.18,010188.14,010188.36,010188.30
Lot NumberP30I010 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010188.18,010188.14,010188.36,010214.14,010188.30
Lot NumberQ30H071 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010188.18,010188.14,010188.36,010214.14,010188.30
Lot NumberP30I011 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010188.18,010188.14,010188.36,010214.14,010188.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium ingot, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.